Cole-Parmer Tensiometer, Capillary Surface Tension Apparatus
Representative image only.
Cole-Parmer Item # WW-59780-90
  • Quickly and easily determine surface tension based on height of liquid in capillary tube
  • Clear, easy-to-read borosilicate glass capillary tube
  • Included rubber stopper centers glass capillary tube in the outer cylinder
  • 2 year warranty
₹53,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTensiometer, Capillary Surface Tension Apparatus
  • Warranty2 year
Two year warranty
Key Features
  • Quickly and easily determine surface tension based on height of liquid in capillary tube
  • Clear, easy-to-read borosilicate glass capillary tube
  • Included rubber stopper centers glass capillary tube in the outer cylinder
  • 2 year warranty
More About this Item
Apparatus is comprised of an engraved 250-mm borosilicate glass capillary tube (graduated from 0 to 10 cm in 1-mm divisions) and an outer tube. Capillary tube is positioned in the center of the outer tube by means of a rubber stopper. Outer tube accommodates a pipette bulb to expel air.