DWK Life Sciences (Kimble) Kimax Reflux condenser, cold finger, 14/20 joint
Representative image only.
DWK Life Sciences (Kimble) Mfr # 283000-0000 – Item # WW-34705-86
  • 1 year warranty
PRODUCT OPTIONS
₹57,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Condenser TypeReflux
  • Coil TypeCold Finger
  • Condenser Joint Size14/20
  • Hose Connection1
  • Coil Length (mm)100
  • Coil Length (in)3.93
  • Approx Condensing Area (cm²)72
  • Approximate Height (mm)200
  • Approximate Height (in)7.87
  • DescriptionReflux condenser, cold finger, 14/20 joint
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Dewar condensers are ideal for liquid nitrogen, dry ice/acetone or ice/alcohol mixtures. Provide rapid condensation of volatile components.
Reflux condensers are complete with an internal coil-type cold finger. outer joint at the top, inner drip joint at the bottom and two hose connectors at the top for water inlet and outlet.