Kestrel 0820 (GREEN) Pocket Wind Meter, LCD Display
Representative image only.
Kestrel Mfr # 0810 (BLUE) – Item # WW-10382-09
Measure air velocity and other weather parameters accurately and easily
  • Wide range of field selectable units
  • Large easy-to-read display
  • Waterproof IP67-rated casing
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Wind Speed (ft/min)7874
  • Min Wind Speed (ft/min)118
  • Wind Speed Resolution1 ft/min
  • Display Type3.5-digit LCD
  • Battery TypeCR2032 battery
  • DescriptionPocket Wind Meter, LCD Display
  • Warranty5 years
5 Year WarrantyConformite EuropeenneIP 67
Key Features
  • Wide range of field selectable units
  • Large easy-to-read display
  • Waterproof IP67-rated casing
  • 5 year warranty
More About this Item
This convenient handheld meter will measure wind speed and other weather parameters with field selectable units including:
  • Knots (KT)
  • Meters per second (M/S)
  • Kilometers per hour (KM/H)
  • Miles per hour (MPH)
  • Feet per minute (FPM)
  • Beaufort force("B")
Kestrel 1000 - Pocket Wind Meter Features
  • Current Wind Speed
  • Max Wind Gust
  • Average Wind Speed
A number of accessories are available including a portable vane mount and a tripod for longer readings, and a replacement impeller.