Kestrel 0804USB USB Interface Cradle for 4000 Series Handheld Meters
Representative image only.
General Tools & Instruments Mfr # 0804USB – Item # WW-10382-08
  • Download data to a computer via USB interface
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUSB Interface Cradle for 4000 Series Handheld Meters
  • Warranty5 years
5 Year WarrantyUL Listed to Canadian safety standards.UL Listed.
Key Features
  • Download data to a computer via USB interface
  • 5 year warranty
More About this Item
We offer a diverse range of handheld weather instrument accessories to maximize the functionality and flexibility of your weather instruments.
Use the full range of Kestrel handheld instrument accessories to protect, hold, and interface the meters to a computer. Let us know if there is a feature you wish to explore for which there is no accessory listed and we will see if it is available.
Take advantage of the wide range of calibration services that are also available to keep your instruments at peak performance and accuracy.
A valuable accessory used with any non-Bluetooth model in the Kestrel 4000 family. With this optional computer interface, data can be uploaded for long-term storage, in-depth analysis and detailed charting.