Kanomax Airflow Transducer Probe, Omni, Spherical, Tipped, 4" L; No Cable
Representative image only.
Kanomax Mfr # 0965-00 – Item # WW-10006-62
Work with the Kanomax airflow transducers
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe length (in)4
  • DescriptionAirflow Transducer Probe, Omni, Spherical, Tipped, 4\" L; No Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer