Kanomax Airflow Transducer Probe, Omni, Mini Bent, 2.125 "L, 100 cm Cable
Representative image only.
Kanomax Mfr # 0965-04 – Item # WW-10006-65
Work with the Kanomax airflow transducers
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe length (in)2.125
  • DescriptionAirflow Transducer Probe, Omni, Mini Bent, 2.125 \"L, 100 cm Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer