Kanomax Airflow Measurement Probe for Velocity, Temperature, and Humidity; Omnidirectional
Representative image only.
Kanomax Mfr # 6533-2G – Item # WW-10007-18
Research-grade accuracy for airflow measurement—ideal for laboratory, IAQ, and critical environment monitoring
  • Smart probe technology lets you change probes without recalibrating the main unit—order meter separately to create your system
  • 1 year warranty
PRODUCT OPTIONS
₹304,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-4
  • Max Temperature (° F)158
  • Min Temperature (° C)-20
  • Max Temperature (° C)70
  • Max RH (%)98
  • Min RH (%)2
  • DescriptionAirflow Measurement Probe for Velocity, Temperature, and Humidity; Omnidirectional
  • Warranty1 year
1 Year Warranty
Key Features
  • Smart probe technology lets you change probes without recalibrating the main unit—order meter separately to create your system
  • 1 year warranty
More About this Item
Unidirectional probes are economical and great for applications where the direction of the airflow is known. Omnidirectional probes can sense airflow from all directions—perfect for turbulent airflow applications.