Justrite 7110100 Steel Safety Can for Flammables, Type I, Flame Arrester, 1 Gal
Representative image only.
Justrite Mfr # 7110100 – Item # WW-81791-41
Safely contain hazardous liquids and control vapors
  • A necessary part of compliance for safety in the workplace
  • Reduce risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations
  • FM approved
  • UL/ULC listed
  • TÜV certified
  • OSHA and NFPA compliant
  • 10 year warranty
PRODUCT OPTIONS
₹13,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel
  • Capacity (Gal)1
  • Capacity (Liters)3.79
  • Exterior Height (in)11
  • Exterior Diameter (in)9.5
  • Exterior Height (cm)27.94
  • Exterior Diameter (cm)24.13
  • ComplianceNFPA, OSHA
  • ApprovalFM, UL, cUL
  • DescriptionSafety Can for Flammables, Type I, 1 gallon
  • Warranty10 Years
10 Year WarrantyFM Approved
Key Features
  • A necessary part of compliance for safety in the workplace
  • Reduce risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations
  • FM approved
  • UL/ULC listed
  • TÜV certified
  • OSHA and NFPA compliant
  • 10 year warranty
More About this Item
All cans include a 3 1/2” built-in, stainless-steel flame arrester to prevent flashback ignition. Self-closing, spring loaded lid automatically vents at 3-5 psi (0.2 and 0.35 bar) to fight rupture or explosion. Reinforcing ribs resist denting or implosion. Steel construction with powder coat paint resists corrosion and improves wear and tear. A large ID zone allows you to label content of the can to avoid misuse
Type I safety cans feature one opening that serves for both filling and pouring. Leakproof lid protects vapors from escaping and guards against spills.