Jonard Industries GD-50 Precision Dynamometer; 60-500 g
Representative image only.
Jonard Industries Mfr # GD-50 – Item # WW-93955-14
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (g)500
  • DisplayAnalog dial with rotational tare ring
  • DescriptionPrecision Dynamometer; 60-500 g
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Measurement is extremely simple. Just place point of gauge arm perpendicular to the force to be measured. Reading can be taken in both directions- clockwise and counter-clockwise.
All Gauges Equipped With Maximum Reading Pointer
Maximum reading pointer moves with indicator when testing. It will remain at maximum force measured, enabling user to take reading afterward. No need to look at dial hand while measuring, thus avoiding reading errors.
Gauges Offer Extreme Precision And Durability
The measuring springs are made of hard-tempered annealed beryllium copper. This alloy provides outstanding flexibility with minimum effort. Dial is protected by unbreakable glass. Packaged in rigid, fitted plastic case to protect instrument.
All gauges equipped with maximum reading pointer…
(Reading + Maximum Scale Reading)
APPLICATIONS
  • Spring tension of all relay contacts and switches
  • Starting torque or motors, clock movements, etc.
  • Pressure of brushes in electro-mechanical devices, phono tone arms, needles, etc.