Jenway 035 262 TrayCell Cuvette for Ultra-Micro Sample Volumes
Representative image only.
Jenway Item # WW-83070-29
The TrayCell is a fibre optic cuvette which can be used with as little as 0.7 µl of sample
  • Ideal for DNA, RNA and protein measurements
  • Sample concentration range from 25 to 4250 µg/ml
  • Sample is simply wiped away after the measurements, making the TrayCell quick and easy to use
  • Prevents dilution and pipetting errors
  • Fits all standard 10 x 10 mm cuvette holders and micro-cuvette holders
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Min Capacity (MicroL)0.7
  • Max Capacity (MicroL)5
  • DescriptionTrayCell Cuvette for Ultra-Micro Sample Volumes
1 Year Warranty
Includes
Caps for 1 mm and 0.2 mm path length operation.
Key Features
  • Ideal for DNA, RNA and protein measurements
  • Sample concentration range from 25 to 4250 µg/ml
  • Sample is simply wiped away after the measurements, making the TrayCell quick and easy to use
  • Prevents dilution and pipetting errors
  • Fits all standard 10 x 10 mm cuvette holders and micro-cuvette holders
  • 1 year warranty
More About this Item
The cuvette is supplied with two caps which give path lengths of either 1 mm or 0.2 mm, thus creating a "virtual dilution" of 1:10 or 1:50 of the sample when compared to a measurement with a standard 10 mm cuvette. Additional or replacement caps can also be purchased separately.
Also for applications of very low or very high DNA concentrations, we can offer two addition caps to provide path lengths of 2 mm or 0.1 mm creating dilutions of 1:5 or 1:100.
Compatible with Genova Bio, Genova Plus, 6705 and 6715 spectrophotometers.