Jenway DNACON Solution Calibration Standard for Micro-Volume Spectrophotometer
Representative image only.
Jenway Item # WW-99959-77
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
  • Reliable, stable, and NIST traceable reference material for the validation of nucleic acid purity measurements using the 260/280 nm ratio method
  • Reference solution consists of a synthetic material whose spectral characteristics closely mimic those of DNA, permanently heat-sealed in a far-UV quartz cell
  • DNACON gives a reliable direct indication of the quality of the overall measurement process
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialDNACON 260/280 ratio solution
  • Compatible WithJenway Genova Nano Spectrophotometer
  • DescriptionDNACON Solution Calibration Standard for Micro-Volume Spectrophotometer
Key Features
  • Expand analysis capabilities or make testing easier
  • Reliable, stable, and NIST traceable reference material for the validation of nucleic acid purity measurements using the 260/280 nm ratio method
  • Reference solution consists of a synthetic material whose spectral characteristics closely mimic those of DNA, permanently heat-sealed in a far-UV quartz cell
  • DNACON gives a reliable direct indication of the quality of the overall measurement process
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
This Reference Material can be used for quality control and assurance of nucleic acid purity measurements using the 260/280 nm ratio method. It is also available packaged for use with dedicated microvolume instruments.