Jenway 027 900 Pure Water Conductivity Probe with ATC, for Benchtop Meters
Representative image only.
Jenway Item # WW-99959-74
Cells for conductivity measurement
  • For use with benchtop and portable meters
PRODUCT OPTIONS
₹60,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantNominal 0.08
  • Body Material316 stainless steel, PEEK, Viton, PVC
  • Min temperature (° C)0
  • Max temperature (° C)50
  • Use WithJenway meters 4510, 4520 and 3540
  • Diameter (mm)12.7
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionPure Water Conductivity Probe with ATC, for Benchtop Meters
Key Features
  • For use with benchtop and portable meters
More About this Item
Compatible with Jenway meters.