International Products Corp Z-0790 Zymit Pro Enzyme Cleaner, 215 KG (197 L)
Representative image only.
International Products Corp Mfr # Z-0790 – Item # WW-18100-19
Biodegradable enzyme and surfactant cleaner that breaks down and removes proteinaceous soils.
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Dilution1:100 to 2:100, pH 7.5
  • ApplicationsBlood, Emulsifiers, Fingerprints, Fragrances, Gels, Proteins
  • Description Z-0790 ZYMIT PRO ENZYME CLEANER 215 KG NET WT PLASTIC DRUM
More About this Item
It is an effective filter membrane cleaner, especially when used with Micro-90 or Micro A07 in consecutive cleaning steps.