This item is now discontinued. We recommend the following as suggested replacements: WW-18100-23
International Products Corp M-1052 Micro Green Clean Biodegradable Cleaner, 20.5 KG (19 L)
Representative image only.
International Products Corp Mfr # M-1052 – Item # WW-18100-09
A free-rinsing, industrial strength, biodegradable hard surface cleaner designed for use in a wide range of applications.
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Dilution1:100 to 2:100, pH 9.8
  • ApplicationsGels, Emulsifiers, Oil, Titanium Doixides, Dyes, Silicaons, Fragrances, Petrolatum, Paraffins
  • Description M-1052 MICRO GREEN CLEAN BIODEGRADABLE CLEANER 20.5 KG NET WT CONTAINER
More About this Item
Excellent for cleaning metals, ceramics, medical instruments, food-processing equipment, filter membranes, and other surfaces. Typical pH: 9.5.