InnoCal NIST-Traceable Calibration; RTD Probe (Probe Only)
Representative image only.
InnoCal Item # WW-17001-04
NIST-traceable calibrations for temperature probes
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
  • Four test points across range of instrument
  • Meter: –200 to 1000°C (–328 to 1832°F); Probe and System: –80 to 1000°C (–112 to 1832°F)
  • Actual range is dependent on type of probe
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeTemperature
  • Probe Only CalibrationYes
  • Number Of Calibration Test Points4 points
  • Calibration Test PointsITS-90 points across the range unless specfied
  • Calibration Test Range-196 to 1300 C
  • DescriptionNIST-Traceable Calibration; RTD Probe (Probe Only)
  • Warranty1 year
1 Year Warranty
Key Features
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
  • Four test points across range of instrument
  • Meter: –200 to 1000°C (–328 to 1832°F); Probe and System: –80 to 1000°C (–112 to 1832°F)
  • Actual range is dependent on type of probe
  • 1 year warranty
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
InnoCal's Metrology Laboratory is accredited to ISO/IEC Guide 25 by the American Association for Laboratory Accreditation (A2LA); certificate number 1746.01. This accreditation is your assurance that one of the foremost outside auditing agencies approves of our internal laboratory policies and procedures.