InnoCal NIST-Traceable Calibration; Load Cell (5 Points), Up to 1000 lbs
Representative image only.
InnoCal Item # WW-17101-38
NIST-traceable calibrations for force gauges, load cells, and scales of all types
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
  • 1 year warranty
PRODUCT OPTIONS
₹27,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionNIST-Traceable Calibration; Load Cell (5 Points), Up to 1000 lbs
  • Warranty1 year
1 Year Warranty
Key Features
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
  • 1 year warranty
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.