InnoCal NIST-Traceable Calibration, Conductivity/Resistivity Meter
Representative image only.
InnoCal Item # WW-17090-30
NIST-traceable calibrations for multimeters, calibrators, oscilloscopes, and more!
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a conductivity/resistivity meter
PRODUCT OPTIONS
₹18,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeElectrical
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsCalibrated per manufacturer's procedure
  • DescriptionNIST-Traceable Calibration; Conductivity/Resistivity Meter
NIST Traceable Calibration
Key Features
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a conductivity/resistivity meter
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
Calibrations on conductivity and resistivity meters are performed using multi-function calibrators, standards-grade reference multimeters, and precision resistors per manufacturer’s procedures. Measurements can be taken to 1100 Mega Ohms and 1000 milliSiemens.
Meters without a detachable probe are tested with standard reference materials from an ISO Guide 34 accredited manufacturer. Measurements are performed per manufacturer’s procedures up to 12,880 microSiemens.