Imada RS-A Type A Reference Specimen Test Block
Representative image only.
Imada Mfr # RS-A – Item # WW-59792-43
Optimize performance of your Imada durometer gauges
  • Compatible with the Imada durometer gauges
  • 2 year warranty
PRODUCT OPTIONS
₹49,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionType A Reference Specimen Test Block
  • Warranty2 year
Two year warranty
Key Features
  • Compatible with the Imada durometer gauges
  • 2 year warranty
More About this Item
Choose from a wide range of accessories and replacement parts for your Imada durometer gauges.
Reference Specimens
These test blocks are convenient references to the durometer’s operating condition. For type A (70 and 50 points).