Imada FB-30N Analog Mechanical Force Gauge, 30 N
Representative image only.
Imada Mfr # FB-30N – Item # WW-93949-90
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Newtons)30
  • Accuracy0.3% FS
  • DisplayAnalog dial with rotational tare ring
  • DescriptionAnalog Mechanical Force Gauge, 30 N
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Simple, rugged, virtually maintenance free
  • Retains a high degree of accuracy for extended periods
  • Ergonomic, heavy-duty metal construction
  • No batteries needed
  • Intrinsically safe in hazardous environments
  • Capacity up to 100 lbs. (push and pull)
  • Overload capacity 165 lbs on all ranges
  • Real time and peak mode
  • Tare ring
  • Full set of accessories and carrying case included
  • Reverse display units are available