Imada E-5000 Constant Load Stand For Durometer Type D
Representative image only.
Imada Mfr # E-5000 – Item # WW-59792-33
Optimize performance of your Imada durometer gauges
  • Compatible with the Imada durometer gauges
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionConstant Load Stand For Durometer Type D
  • Warranty2 year
Two year warranty
Key Features
  • Compatible with the Imada durometer gauges
  • 2 year warranty
More About this Item
Choose from a wide range of accessories and replacement parts for your Imada durometer gauges.
Constant Load Stand with 5000g weight
Series 'e' constant load stand eliminates the load variables which occur when testing samples with a durometer by hand. The test load the durometer applies to the test sample is kept constant with a standard 5,000g weight.
Durometer is sold separately.