IKA MT 40.10 Disposable Grinding Chamber, 40 mL; 10/pk
Representative image only.
IKA Mfr # 0004425000 – Item # WW-28711-15
Eliminate contamination with disposable grinding chamber
  • Easily view and clean with transparent, disposable grinding chamber
  • Durable construction suitable for grinding material
PRODUCT OPTIONS
₹11,807.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Sample Capacity (mL)40
  • MaterialPolypropylene chamber with stainless steel blade
  • DescriptionDisposable Grinding Chamber, 40 mL; 10/pk
Key Features
  • Easily view and clean with transparent, disposable grinding chamber
  • Durable construction suitable for grinding material
More About this Item
Choose from a wide range of accessories for the IKA Benchtop Tube Mill Control Mills, items 28710-88 and 28710-89. Mills are sold separately.