IKA Labworldsoft® 6 Starter Software, Limited to 3 Devices
Representative image only.
IKA Mfr # 0020019397 – Item # WW-28711-52
Laboratory analysis software that is cross-functional between up to 40 manufacturers
  • Has a library of over 700 devices
  • Customized device integration is possible
  • Rapidly structure complex applications with a large number of functional modules
  • Export data to 3 different file types as needed
  • All data is safely located on your own network
  • Limit of 3 devices
PRODUCT OPTIONS
₹544,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Operating PlatformWindows
  • Operating System VersionWindows 7, 8, or 10
  • DescriptionStarter Software, Limited to 3 Devices
Two year warranty
Includes
License activation key.
Key Features
  • Has a library of over 700 devices
  • Customized device integration is possible
  • Rapidly structure complex applications with a large number of functional modules
  • Export data to 3 different file types as needed
  • All data is safely located on your own network
  • Limit of 3 devices
More About this Item
This software allows users to pair IKA devices with any number of others in their laboratory. With the removal of company-specific software, anything is possible with this program. It can capture multiple physical parameters such as pH values, conductivity, temperature, torque, weight, or flow rate simultaneously. Functional modules allow for reproduction of any experiment and configuration of multiple values into a single test.
Labworldsoft 6 Starter allows the configuration of up to 3 devices. Labworldsoft 6 Pro allows access to the entire device library, configuring as many devices as you would like.