IKA 8039500 Sawtooth dispersing element for T-50 homogenizer, 45 G ST
Representative image only.
IKA Mfr # 0008039500 – Item # WW-04738-82
Quickly homogenize tissues and solids in batches up to 20 L
  • Homogenation sample volume of 0.5- to 20-L
  • Ultimate fineness of 40 μm
  • 2 year warranty
₹361,701.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSawtooth dispersing element for T-50 homogenizer, 45 G ST
  • Warranty2 year
Two year warranty
Key Features
  • Homogenation sample volume of 0.5- to 20-L
  • Ultimate fineness of 40 μm
  • 2 year warranty
More About this Item
Compatible for use with IKA® T50 ULTRA-TURRAX® Heavy-Duty Homogenizers 04738-30 and -35. Homogenizer sold separately.