IKA 0020023170 Wing Style Beater, Stainless Steel, 400 mL
Representative image only.
IKA Mfr # 0020023170 – Item # WW-28711-61
Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill
  • Compatible with IKA grinder mill 28711-53
  • Suitable for most brittle materials up to Mohs hardness 5
  • Compatible with IKA MultiDrive MI 400 (28711-56)
SPECS & DESCRIPTION
Specifications & Description
  • Sample Capacity (mL)400
  • MaterialStainless steel
  • DescriptionStandard Beater, Stainless Steel
Two year warranty
Key Features
  • Compatible with IKA grinder mill 28711-53
  • Suitable for most brittle materials up to Mohs hardness 5
  • Compatible with IKA MultiDrive MI 400 (28711-56)
More About this Item
Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill