Idex XP-308 Flangeless Fitting, Short Head, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # XP-308 – Item # WW-02020-93
Short head is ideal for tight-speace applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Black acetal
  • 1/EA
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorBlack
  • Warranty1 year
1 Year Warranty
Includes
Yellow ETFE ferrule 02019-82.
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Black acetal
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.