Idex VHP-900-01X VHP Ferrule, SS, 1/32" OD Tubing, M4 Coned; 10/PK
Representative image only.
Idex Mfr # VHP-900-01X – Item # WW-02022-84
Ferrules for use with VHP fittings
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Replacement ferrule for VHP SS fittings 02022-82 and -83
  • Max pressure 30,000 psi (2070 bar)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSS
  • Compatible WithReplacement ferrule for VHP SS fittings 02022-82 and -83
  • DescriptionVHP Ferrule, SS, 1/32\" OD Tubing, M4 Coned; 10/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Replacement ferrule for VHP SS fittings 02022-82 and -83
  • Max pressure 30,000 psi (2070 bar)
  • 1 year warranty
More About this Item
These fittings ensure reliable connections and performance for your instruments.