Idex V-241 Manual Selection Valve, Black, Bulkhead, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA
Representative image only.
Idex Mfr # V-241 – Item # WW-02014-90
Allow operator control of low-use application valves
  • Enable connections among multiple fluid systems 6-position, 7-port configuration
  • Black
  • Bulkhead
  • 1/4-28 flat-bottom ports for 1/16" OD tubing
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4-28 flat bottom
  • Inlet Port1/4-28 flat bottom
  • Max Pressure (PSI)1000
  • Max Pressure (bar)69
  • Wetted MaterialsPolyimide/PTFE
  • Tubing OD (in)0.0625
  • DescriptionManual Selection Valve, Bulkhead, 6-Position, 7-Port, 1/16\" OD Tubing; 1/EA
1 Month Warranty
Includes
Six flangeless natural PEEK fitting with short head for 1/16" OD tubing.
Key Features
  • Enable connections among multiple fluid systems 6-position, 7-port configuration
  • Black
  • Bulkhead
  • 1/4-28 flat-bottom ports for 1/16" OD tubing
  • 1 month warranty
More About this Item
Manual valves serve well in applications that require operator control or involve lesser frequency of use. Selection valves enable connection to one column for another without the need to manually disconnect the plumbing. In sequencing or diagnostic applications, the valve alternates between the sample or reagent streams.