Idex P-405 Flanged Fitting, 5/16" Square, Green Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-405 – Item # WW-02025-59
Excellent for low to moderate pressure applications
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
  • For 1/16" OD tubing
  • Green Delrin® acetal nut
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorGreen
  • DescriptionFlanged Fitting, 5/16\" Square, Green Acetal, 1/16\" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Includes
Stainless steel washer.
Key Features
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
  • For 1/16" OD tubing
  • Green Delrin® acetal nut
  • 1 year warranty
More About this Item
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.