Idex P-296 PEEK Replacement Tubing, Ferrule Assembly for Injection Port Adapters; 1/4-28
Representative image only.
Idex Mfr # P-296 – Item # WW-02032-13
Prevent samples from veering away from the flow path
  • For Use with Any Standard High Pressure Injection Valve
  • Replacement Tubing Ferrule Assembly for Injection Port Adapters For 1/4-28 Flat-Bottom Port Configurations
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Key Features
  • For Use with Any Standard High Pressure Injection Valve
  • Replacement Tubing Ferrule Assembly for Injection Port Adapters For 1/4-28 Flat-Bottom Port Configurations
  • 1 year warranty
More About this Item
These adapters are made so the least amount of sample is wasted.
A. Injection Port Adapters eliminate the potential for additional swept volume to the fluid pathway since the needle butts directly against the connecting tubing. They can be bulkhead mounted or mounted with the insulated mounting bracket assembly 02032-11.
B. Suction Needle Adapters allows for suction-loading of a valve for metal-free loading of a sample loop.