Idex P-258-01B Super Flangeless™ Ferrule, Blue ETFE; 1/EA
Representative image only.
Idex Mfr # P-258-01B – Item # WW-02021-68
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year Warranty
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • 1 year warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Minimum order 25 units.