Idex Manual Switching Valve, Double 3-Way, SS/Vespel®, 2-Position, 6-Port; 1/EA
Representative image only.
Idex Mfr # 7030 – Item # WW-02015-23
Easily switch between two fluid paths without disconnecting the lines
  • Enable connections among multiple fluid systems
  • Double 3-way
  • 10-32 coned ports for 1/16" OD tubing
  • 1 year warranty
PRODUCT OPTIONS
₹156,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port10-32 coned
  • Inlet Port10-32 coned
  • Max Pressure (PSI)7000
  • Max Pressure (bar)483
  • Wetted MaterialsSS/Vespel
  • Tubing OD (in)0.0625
  • Flow PatternDouble three-way, two-position
  • DescriptionManual Switching Valve, Double 3-Way, SS/Vespel®, 2-Position, 6-Port; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Enable connections among multiple fluid systems
  • Double 3-way
  • 10-32 coned ports for 1/16" OD tubing
  • 1 year warranty
More About this Item
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.