Idex M-126 NanoFilter Capsule for M-537/M-538, Red PEEK, 1 µm Titanium Frit; 2/PK
Representative image only.
Idex Mfr # M-126 – Item # WW-42711-25
Use with solvent MicroFilters to protect your columns from particulates
  • Optimize performance of your solvent MicroFilters
  • Red PEEK body, 1 µm Titanium frit
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPEEK
  • ColorRed
  • Compatible WithM-537/M-538
  • DescriptionNanoFilter Capsule for M-537/M-538, Red PEEK, 1 µm Titanium Frit; 2/PK
1 Month Warranty
Key Features
  • Optimize performance of your solvent MicroFilters
  • Red PEEK body, 1 µm Titanium frit
  • 1 month warranty
More About this Item
Choose from a wide range of accessories including end-fittings, capsules, and more suitable for use with your Idex® inline solvent MicroFilter.