Idex Flangeless Ferrule, White ETFE, 2.5 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-353 – Item # WW-12029-63
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
PRODUCT OPTIONS
₹510.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (mm)2.5
  • ColorWhite
  • Max Operating Pressure (PSI)500
  • DescriptionFlangeless Ferrule, White ETFE, 2.5 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA
Key Features
  • Removable and reusable—saving you money!
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.