Idex Precolumn Filter, Female End, Stainless Steel
Representative image only.
Idex Mfr # A-315-01 – Item # WW-02010-29
Precolumn filters offer secure protection for analytical columns
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16" OD tubing
  • Replaceable stainless steel (SS) and PEEK® frits
  • Applications: Alcohols, aromatics, flavors, free fatty acids, low-polarity pesticides
  • 1 year warranty
PRODUCT OPTIONS
₹7,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialStainless Steel
  • Housing MaterialStainless Steel
  • Pore Size (µm)2
  • ConnectionsFemale End
  • DescriptionPrecolumn Filter, Female End; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16" OD tubing
  • Replaceable stainless steel (SS) and PEEK® frits
  • Applications: Alcohols, aromatics, flavors, free fatty acids, low-polarity pesticides
  • 1 year warranty
More About this Item
Economical precolumn filters are designed to protect columns by filtering out particulate matter originating from the sample or from rotor seal wear. Filters feature 10-32 coned ports compatible with 1/16" outer diameter tubing.