Idex RheBuild® Kit for Valve Models 3725/3725i/3725-038/3725i-038; 1/EA
Representative image only.
Idex Mfr # 3725-999 – Item # WW-02015-34
Regular maintenance ensures optimal valve performance
  • Optimize performance of your manual chromatography valves
  • Use with valve models 3725/3725i/3725-038/3725i-038
  • 1 year warranty
PRODUCT OPTIONS
₹43,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithValve models 3725/3725i/3725-038/3725i-038
  • DescriptionRheBuild® Kit for Valve Models 3725/3725i/3725-038/3725i-038; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Optimize performance of your manual chromatography valves
  • Use with valve models 3725/3725i/3725-038/3725i-038
  • 1 year warranty
More About this Item
RheBulid® kits contains normal wear parts, tools for disassembly/reassembly, and instructions to maintain your valves.