Idec HW1L-M1F10QD-G-24V Illuminated Push-button Switch, round, green
Representative image only.
Idec Mfr # HW1L-M1F10QD-G-24V – Item # WW-65518-40
22 mm industrial plastic bezel devices use slow-make, double break contacts for switching of heavier loads
  • Tamperproof mounting and construction
  • Slow make, double break, self cleaning 10 A contacts make a longer-lasting product
  • 600 V/10 A contacts with a wide operating range from 5 mA at 3 V AC/DC to 10 A at 120 VAC
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Load Rating600 VAC
  • Control Voltage3 VAC/DC (at 5 mA) to 120 VAC (at 10 A)
  • Max Current (amps)10
  • DescriptionIlluminated Push-button Switch, round, green
  • Warranty1 year
1 Year Warranty
Key Features
  • Tamperproof mounting and construction
  • Slow make, double break, self cleaning 10 A contacts make a longer-lasting product
  • 600 V/10 A contacts with a wide operating range from 5 mA at 3 V AC/DC to 10 A at 120 VAC
  • 1 year warranty
More About this Item
Fully assembled switches and pilot devices are IP65 rated and designed for washdown applications in pharmaceutical, food and beverage, and general process areas. Select from illuminated and non-illuminated pushbuttons, pilot devices, and selector switches. These IEC style devices feature tamperproof mounting and construction, modular stackable contact blocks with IP20 fingersafe screw terminals, and pure silver contacts with a self cleaning action.
Safety and ease of use are evidenced by the simple mounting procedure. Mechanically keyed safety lever lock prevents contact separation in heavy vibration applications. Contact blocks are color coded and are internationally recognized.