HTL 5124 Discovery Comfort DV8-300 Multichannel Pipettor with Gray Shaft, Green, 8-Channel; 50 to 300 µL
Representative image only.
Mfr # 5124 – Item # WW-24551-66
Comfort, safety, precision and value make this variable-volume pipette series quite a discovery
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)300
  • Min Volume (µL)50
  • Accuracy (%)1
  • Precision (%)0.6
  • Number Of Channels8
  • Tip EjectorYes
  • Color CodeGreen
  • DescriptionDiscovery Comfort DV8-300 Multichannel Pipettor with Gray Shaft, Green, 8-Channel; 50 to 300 µL
  • Warranty3 year
3 Year Warranty
Key Features
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision. Multichannel models feature a revolutionary suspension system which allows the shafts to retract slightly when they are pressed against a row of pipette tips.