Horiba SR-305 Solvent Reclaimer for S-316 Extraction Solvent
Representative image only.
Horiba Mfr # 3014102236 – Item # WW-82900-20
Save money on solvent costs
  • Reduce overall volume of solvents used and discarded
  • Solvent reclaimer solution
PRODUCT OPTIONS
₹464,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSolvent reclaimer SR-305, for recycling Oil Content Analyzer extractant
Key Features
  • Reduce overall volume of solvents used and discarded
  • Solvent reclaimer solution
More About this Item
Designed especially for reclaiming S-316 solvent, this unit features a double column of activated carbon and activated aluminum. It has a large filtering capacity, is easy to operate, and requires no electricity. Highly efficient, the SR-305 can reduce your per test solvent cost.