Honeywell Rig Dog™ Waterproof Impact ANSI A6 Cut-Resistant Glove, Slip-On Cuff, 8/M
Representative image only.
Honeywell Mfr # 42-615BL/8M – Item # WW-86326-57
Impact-resistant gloves protect hands in tough wet environments
  • Enhanced grip and dexterity ideal for wet applications ANSI A6 (ISO level F) cut resistance; impact protection level 2 Rapid sweat absorption aids comfort for extended wear Slip-on cuff for easy donning
  • Size 8/M
SPECS & DESCRIPTION
Specifications & Description
  • Size8/M
  • Length (in)10
  • Length (cm)25.4
  • ColorBlack/Blue
  • ApprovalANSI A6 (ISO level F) cut resistance, impact protection level 5
  • Max Temperature (° F)86
  • Max Temperature (° C)30
  • DescriptionWaterproof Impact ANSI A6 Cut-Resistant Glove, Slip-On Cuff, 8/M
Key Features
  • Enhanced grip and dexterity ideal for wet applications ANSI A6 (ISO level F) cut resistance; impact protection level 2 Rapid sweat absorption aids comfort for extended wear Slip-on cuff for easy donning
  • Size 8/M
More About this Item
Smartly-designed for safety, Rig Dog™ waterproof gloves from Honeywell feature ANSI A6 cut protection. CoreNest™ technology with reinforced thermoplastic rubber maximizes impact protection for the back of the hand, including fingertips, knuckles and the thumb crotch.
Minimum stitching on palm provides low oil permeability for comfort and durability.