Honeywell 32-001100-0000 Bacteria-inhibiting eyewash preservative
Representative image only.
Honeywell Mfr # 32-001100-0000 – Item # WW-06796-66
NOT AVAILABLE IN CANADA
  • Solution improves the performance and safety of self-contained eyewash stations
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBacteria-inhibiting eyewash preservative
Key Features
  • Solution improves the performance and safety of self-contained eyewash stations
More About this Item
De-fend® solution (06796-66) inhibits the growth of harmful bacteria in your self-contained eyewash station. One 8-oz bottle is ideal for use in 14- to 16-gallon stations. For smaller stations, proper concentration is ½ oz of preservative for every gallon of potable water. Bottle is graduated to assist in mixing.
Solution has a two-year shelf life and is stamped with lot number and date code.