Cole-Parmer Essentials Hollow Polypropylene (PP) Ball Bath Covers, 38 mm; 1000/Pk
Representative image only.
Cole-Parmer Essentials Item # WW-06821-08
Reduce vapor and heat loss by 75% with one layer of balls
  • 3 month warranty
PRODUCT OPTIONS
₹74,800.00INR / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHollow Polypropylene (PP) Ball Bath Covers, 38 mm; 1000/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.