High-Tech Conversions NT56-99 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 9" x 9", 3000/CS
Representative image only.
High-Tech Conversions Mfr # NT56-99 – Item # WW-15981-21
Lint-free, versatile, and highly effective
  • Converted in an ISO Class 6 cleanroom to maintain certification Polyester and cellulose blend is tough, absorbent, and abrasion-resistant Economical choice for general-purpose wiping
  • 2 year warranty
PRODUCT OPTIONS
₹44,700.00INR / Case of 3000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester / Cellulose Blend
  • ColorWhite
  • ComplianceISO Class 6
  • Width (in)9
  • Length (in)9
  • Width (cm)23
  • Length (cm)23
  • DescriptionCleanroom Wipes, Non-Woven, Lint-Free, Polyester / Cellulose Blend, 9\" x 9\", 3000/CS
  • Warranty2 year
Two year warranty
Key Features
  • Converted in an ISO Class 6 cleanroom to maintain certification Polyester and cellulose blend is tough, absorbent, and abrasion-resistant Economical choice for general-purpose wiping
  • 2 year warranty
More About this Item
Use these lightweight cleanroom wipes for light- to moderate-duty cleaning applications. The blend of virgin polyester and cellulose forms a strong fabric virtually free of particle generation. The ISO 6-certified fabric is also converted in a cleanroom facility minimizing exposure to pollutants.