High-Tech Conversions Critical Task Wipe, 15 x 16.6? Single-Ply Cellulose; 140 Wipes/Box
Representative image only.
High-Tech Conversions Mfr # Techwipe-XL – Item # WW-78902-96
Eco-friendly lab wipes made from 100% recycled cellulose
  • Ideal for optics and other sensitive surfaces
  • Perfect wipe for large surface areas
₹2,000.00INR / Pkg of 140
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSingle-ply cellulose
  • Width (in)15
  • Length (in)16 19/32
  • DescriptionFriendly Green Critical Task Wipe, 15″ x 16.6″ Single-Ply Cellulose; 140 Wipes/Box
Key Features
  • Ideal for optics and other sensitive surfaces
  • Perfect wipe for large surface areas
More About this Item
Made from virgin cellulose, these lab wipes are excellent for your light-duty applications, will not scratch, and can be used in a variety of industries. The wipes are made of single-ply, very absorbent, and soft material making them ideal for cleaning optics and sensitive instruments and equipment in your lab facility. Since they are low linting and low in chemical extractables, they’ll leave surfaces extremely clean.
The 15? x 16.6? (38 x42 cm) wipes are packaged 140 per box.