High Five General Purpose Latex Gloves, Medium, Powdered, 100/box
Representative image only.
High Five Mfr # L492 – Item # WW-86232-36
Elasticity provides exceptional glove dexterity
  • Textured for a more secure grip
  • Disposable
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)5.5
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • GripTextured
  • CuffBeaded
  • SterileNo
  • PowderedYes
  • DescriptionGeneral Purpose Latex Gloves, Medium, Powdered, 100/box
Key Features
  • Textured for a more secure grip
  • Disposable
More About this Item
General-purpose latex gloves are a great choice for food processing and other industrial areas. Glove surface is textured for a more secure grip; beaded cuff. Disposable; for single-use only. Ambidextrous.
These gloves are powdered for easy donning.