Ashcroft 1305D Deadweight Tester Kit, 5000 psi Maximum
Representative image only.
Ashcroft Mfr # 1305D-50 – Item # WW-68840-30
Primary pressure standard accurate to ±0.1% of reading
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure5000 psi
  • Min Pressure15 psi
  • Piston / Cylinder Material400-series stainless steel
  • Calibration MediumSAE 20 automotive or machine oil (approximately 1-1/2 pints required; not included)
  • DescriptionDeadweight Tester Kit, 5000 psi Maximum
  • Warranty1 Year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • 1 year warranty
More About this Item
These self-contained tester kits have everything you need for testing, setting, calibrating, and repairing your high-accuracy pressure instruments—except operating fluid. Each kit consists of a two-stage hydraulic pump, two interchangeable piston assemblies (high- and low-pressure), a manifold with connectors, and an assortment of nonmagnetic, die cast zinc alloy weights. Precision-tooled weights are premeasured, marked with pressure values for each piston and come with a separate case.
Use the high-pressure piston for calibration pressures at maximum psi with a 75 psi resolution (see table at below); use the low-pressure piston to achieve 1/5 of the maximum pressure with a 15 psi resolution.
During operation, the manifold is pressurized until the weight assembly reaches a designated mark. A displacement valve lets you vary the pressure without using the pump handle or release valve. Once the correct pressure is achieved, simply reposition the needle on your gauge to the corresponding value.
Max psi in the table is with a 75 psi resolution using the high-pressure piston.