Haws 8356WCSM Pull-Down Eyewash with Shower
Representative image only.
Haws Mfr # 8356WCSM – Item # WW-06767-52
Inverted flow of eyewash sweeps contaminants away from inner eye and nasal cavity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Flow Rate (GPM)3.7
  • Shower Flow Rate (GPM)20
  • Bowl MaterialStainless steel
  • Shower Head MaterialStainless steel
  • MountWall Mount
  • Outlet Connections2\" NPT(F)
  • Inlet Connection1\" NPT(F)
  • ComplianceANSI Z358.1-2009
  • DescriptionPull-Down Eyewash with Shower
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Pull-down eyewash features a unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols used by laboratory research professionals. With other eyewashes, contaminants can travel through the inner corner of the eye to the nasal cavity resulting in further injury. Type 304 stainless steel cabinet is surface mounted for installation wherever you need it. Simply pull down the eywash to activate water flow. Eyewash/shower model features a separate pull-down handle to activate the shower.
Compliance: ANSI Z358.1-2009
Eyewash/shower model features a separate pull-down handle to activate the shower.