Haws 7620 Faucet-Mount Eyewash, stainless steel
Representative image only.
Haws Mfr # 7620 – Item # WW-06767-01
Inverted flow sweeps contaminants away from inner eye and nasal cavity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Fixture MaterialStainless steel
  • MountFaucet Mount
  • Inlet Connection55/64\"-27 (F) for faucet
  • ComplianceANSI Z358.1-2009, CSA B125.1, ASME A112.18.1
  • DescriptionFaucet-Mount Eyewash, stainless steel
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Faucet-mounted eyewash features a unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols. With other eyewashes contaminants can travel through the inner corner of the eye to the nasal cavity, resulting in further injury. Rotate the eyewash to activate — thermostatic protection shuts off the eyewash if the temperature reaches 100°F (37.8°C). Rugged stainless steel finish stands up to industrial applications.
Compliance: ANSI Z358.1-2009, CSA B125.1, and ASME A112.18.1