Haws 7260B-7270B Eyewash Bowl, Wall-mount, ABS Plastic bowl
Representative image only.
Haws Mfr # 7260B-7270B – Item # WW-06767-31
Inverted flow sweeps contaminants away from inner eye and nasal cavity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Flow Rate (GPM)3.7
  • Bowl MaterialABS plastic
  • MountWall Mount
  • Outlet Connections1-1/4\" IPS
  • Inlet Connection1/2\" IPS
  • ComplianceANSI Z358.1-2009
  • DescriptionEyewash Bowl, Wall-mount, ABS Plastic bowl
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Eyewashes feature a unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols. With other eyewashes contaminants can travel through the inner corner of the eye to the nasal cavity resulting in further injury.
Bowls are 11" (27.9 cm) in diameter and include a push handle to start flow. Supply connection is 1/2" IPS; waste connection is 1-1/4" IPS.
Compliance: ANSI Z358.1-2009