Hamilton Syringe Needle with Kel-F Hub, Beveled Tip, 22s G; 6/Pk
Representative image only.
Hamilton Mfr # 90138 – Item # WW-07938-97
Ideal for chromatography
  • Constructed of durable 304 stainless steel
  • Autoclavable
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • 18 month warranty
PRODUCT OPTIONS
₹12,200.00INR / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeKel-F hub (KF) needle
  • Needle Tip StyleSharp, beveled, curved, non-coring
  • Needle Tip Number2
  • Needle Gauge22S
  • Needle Length (in)2
  • Material304 stainless steel
  • DescriptionSyringe Needle with Kel-F Hub, Beveled Tip, 22s G; 6/Pk
  • Warranty18 months
18 Month Warranty
Key Features
  • Constructed of durable 304 stainless steel
  • Autoclavable
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • 18 month warranty
More About this Item
Hamilton® Syringe Needle with Kel-F hub (KF) can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2" / 51 mm.