Hamilton Repeating Syringe Dispenser for 10 µL to 2.5 mL Syringes
Representative image only.
Hamilton Mfr # 83700 – Item # WW-07939-95
Low cost high accuracy accessory for repetitive dispensing
  • Dispense volumes between 0.2 – 50 µL
  • Easily and consistently dispense samples up to 50 times with the push of a button
  • 18 month warranty
₹22,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeRepeating Dispenser
  • Compatible WithHamilton 700 and 1700 series syringes: 10 µL, 25 µL, 50 µL, 100 µL, 250 µL, 500 µL, 1 mL, and 2.5 mL
  • DescriptionRepeating Syringe Dispenser for 10 µL to 2.5 mL Syringes
  • Warranty18 months
18 Month Warranty
Key Features
  • Dispense volumes between 0.2 – 50 µL
  • Easily and consistently dispense samples up to 50 times with the push of a button
  • 18 month warranty
More About this Item
The Hamilton® repeating syringe dispenser is designed to consistently dispense samples up to 50 times with the push of a button. Easily dispense volumes between 0.2 µL to 50 µL. This dispenser works with Hamilton® 700 and 1700 series microliter or Gastight® syringes from 10 µL to 2.5 mL. Use the repeating dispenser with cemented needle, removable needle, luer tip, or PTFE Luer Lock syringes. Syringes sold separately.